O2-tert-butyl O3-ethyl (1S,3S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.2]octane-2,3-dicarboxylate 1 * 1 g
Catalog # PHBLPBWP405-1G
Supplier: Pharmablock (USA)
O2-tert-butyl O3-ethyl (1S,3S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.2]octane-2,3-dicarboxylate 1 * 1 g
Catalog # PHBLPBWP405-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
HFIPA LC-MS 1 * 100 mL
Supplier: Novartis
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific