4-bromo-5-methyl-pyrimidin-2-amine 1 * 1 g
Catalog # PHBLPBU5847-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Petroleum spirit, 60…95 °C, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
n-Pentane, EMSURE® for analysis, Supelco®
Supplier: Merck
Trichloroethylene ≥98%
Supplier: Thermo Fisher Scientific
Water suitable for ion chromatography, Supelco®
Supplier: Merck
Tetramethylammonium chloride, Supelco®
Supplier: Merck
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific