6-amino-5-fluoro-pyridine-3-carbonitrile 1 * 1 g
Catalog # PHBLPBU5227-1G
Supplier: Pharmablock (USA)
6-amino-5-fluoro-pyridine-3-carbonitrile 1 * 1 g
Catalog # PHBLPBU5227-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Sulphuryl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrachloroethylene ≥98%, TECHNICAL
Supplier: VWR Chemicals
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
n-Pentane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dichloromethane 99.9% stabilised for residue analysis, for trace analysis of polyaromatic hydrocarbons
Supplier: Thermo Fisher Scientific
n-Heptane 99.5% for analysis
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.9%
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Carbon disulphide ≥99.9%, Reagent Grade low in benzene
Supplier: Honeywell Chemicals
N-HEXANE 99% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals