2-methyl-6,7-dihydro-5H-indazol-4-one 1 * 1 g
Catalog # PHBLPBU5172-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98.5% (isomers plus ethylbenzene) ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Hydrogen chloride 1.25 mol/l in methanol, Supelco®
Supplier: Merck
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Ethoxyethanol 99%, Reagent Grade
Supplier: Honeywell Chemicals
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
1,2-Dichloroethane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
EAU STÉRILE APYROGÈNE BIDON DE 5 LITRES 1 * 5 L
Supplier: Sorenson Bioscience