2-[(3R)-3-piperidyl]acetic acid hydrochloride 1 * 1 g
Catalog # PHBLPBU5167-1-1G
Supplier: Pharmablock (USA)
2-[(3R)-3-piperidyl]acetic acid hydrochloride 1 * 1 g
Catalog # PHBLPBU5167-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥99.7% (by GC), Puriss., meets analytical specification of Ph.Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific
n-Hexane ≥95% for spectrophotometry
Supplier: Thermo Fisher Scientific
Chloroform ≥99.0% stabilised BP, USP, NF for UV spectrophotometry, for IR spectrophotometry, for HPLC
Supplier: PanReac AppliChem
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific