(3S,6S)-3,6-dimethyl-1,4-dioxane-2,5-dione 1 * 10 g
Catalog # PHBLPBTF730-10G
Supplier: Pharmablock (USA)
(3S,6S)-3,6-dimethyl-1,4-dioxane-2,5-dione 1 * 10 g
Catalog # PHBLPBTF730-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals