3-thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride 1 * 1 g
Catalog # PHBLPBTEN115401-1G
Supplier: Pharmablock (USA)
3-thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride 1 * 1 g
Catalog # PHBLPBTEN115401-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexane, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile for DNA synthesis
Supplier: Merck
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.8% for HPLC
Supplier: Thermo Fisher Scientific