7-bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 1 * 1 g
Catalog # PHBLPBTB205-1G
Supplier: Pharmablock (USA)
7-bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 1 * 1 g
Catalog # PHBLPBTB205-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99.8%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Pyrrolidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Indole for synthesis, Sigma-Aldrich®
Supplier: Merck
Chloroform, extra dry over Molecular Sieve 99.9% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, Supelco®
Supplier: Merck
Water, pure, demineralised
Supplier: Thermo Fisher Scientific
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-Dimethylacetamide 99%, Reagent Grade
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ AMD, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor