5-bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 1 * 1 g
Catalog # PHBLPBTB204-1G
Supplier: Pharmablock (USA)
5-bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 1 * 1 g
Catalog # PHBLPBTB204-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Paraffin, liquid, liquid Ph. Eur.
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
o-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals