N-[1-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-hydroxy-3-(2-methoxyethoxy)tetrahydrofuran-2-yl]-2-oxo-pyrimid in-4-yl]benzamide 1 * 1 g
Catalog # PHBLPBT8305-1G
Supplier: Pharmablock (USA)
N-[1-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-hydroxy-3-(2-methoxyethoxy)tetrahydrofuran-2-yl]-2-oxo-pyrimid in-4-yl]benzamide 1 * 1 g
Catalog # PHBLPBT8305-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals