(S)-13-hydroxy-10,16-bis(2,4,6-triisopropylphenyl)-12,14-dioxa-13phosphapentacyclo[13.8.0.0.0 .0 ]tricosa-1(15),2,4,6,8,10,16 ,18(23),19,21-decaene 13-oxide 1 * 1 g
Catalog # PHBLPBT4874-1G
Supplier: Pharmablock (USA)
(S)-13-hydroxy-10,16-bis(2,4,6-triisopropylphenyl)-12,14-dioxa-13phosphapentacyclo[13.8.0.0.0 .0 ]tricosa-1(15),2,4,6,8,10,16 ,18(23),19,21-decaene 13-oxide 1 * 1 g
Catalog # PHBLPBT4874-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
SIGTECH Mek Solvent 5 L Kanister 1 * 5 L
Supplier: MARKET SOURCE PART PROCESS
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Ethyl acetate ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
Liquid for manometer with sloping tube
Supplier: VWR Chemicals
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals