tert-butyl 4-[(4-formylphenyl)methyl]piperazine-1-carboxylate 1 * 1 g
Catalog # PHBLPBT3725-1G
Supplier: Pharmablock (USA)
tert-butyl 4-[(4-formylphenyl)methyl]piperazine-1-carboxylate 1 * 1 g
Catalog # PHBLPBT3725-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Biosolve Methanol ULC/MS grade, 6*1Liter 1 * 6 L
Supplier: Novartis
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
n-Pentane for analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals