3,5-dibromo-6-methyl-pyrazin-2-amine 1 * 1 g
Catalog # PHBLPBT3580-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Chloroform, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
1,8-Diaminonaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Super-DHB ≥99.0% matrix substance for MALDI, Supelco®
Supplier: Merck
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific