3-bromopyrazine-2-carboxamide 1 * 1 g
Catalog # PHBLPBT3574-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Hydrindantin dihydrate, Supelco®
Supplier: Merck
2-Propanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Fisher Scientific
Toluene 99.85% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific