5-aminopyrazine-2-carboxamide 1 * 1 g
PHBLPBT3571-1G
Frequently Bought Together
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
Cyclohexane, EMPLURA®, Supelco®
: Merck
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals