1-methyl-2H-pyrrol-5-one 1 * 1 g
Catalog # PHBLPBT3555-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,4-Dioxane, anhydrous (max. 0.003% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene, extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Supplier: Thermo Fisher Scientific
Acetone 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific