2-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
PHBLPBT3340-1G
2-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
PHBLPBT3340-1G
Frequently Bought Together
Lithium perchlorate trihydrate 99%, extra pure
: Thermo Fisher Scientific
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran ≥99% stabilised
: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Diphosphorus pentoxide, Supelco®
: Merck
n-Hexane ≥95% for spectrophotometry
: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
2-Butoxyethanol ≥99%
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
Isobutanol 99+% for HPLC
: Thermo Fisher Scientific
Methanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
2-Methylbutane ≥99%, Reagent Grade
: Honeywell Chemicals