SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
methyl 4-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]benzoate 1 * 1 g
PHBLPBT2165-1G
:
Pharmablock (USA)
methyl 4-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]benzoate 1 * 1 g
PHBLPBT2165-1G
:
Pharmablock (USA)
Frequently Bought Together
Methanol : Water (30:70) % (v/v), LiChrosolv®, Supelco®
: Merck
(±)-2-Methyltetrahydrofuran, EMPLURA®
: Merck
N,N-Dimethylacetamide 99.5+% for analysis
: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Cyclopentane for HPLC
: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C, CHROMASOLV™ for pesticide residue analysis, low boiling point hydrogen treated naphtha, Riedel-de Haën™
: Honeywell Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
: VWR Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals