1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)-3-methyloxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione 1 * 10 g
Catalog # PHBLPBSQ8028-10G
Supplier: Pharmablock (USA)
1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)-3-methyloxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione 1 * 10 g
Catalog # PHBLPBSQ8028-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
m-Xylene, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.8% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals