N-[9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-6-oxo-1H-purin-2-yl]-2-methyl-propanamide 1 * 1 g
Catalog # PHBLPBQS409-1G
Supplier: Pharmablock (USA)
N-[9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-6-oxo-1H-purin-2-yl]-2-methyl-propanamide 1 * 1 g
Catalog # PHBLPBQS409-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
2,2,4-Trimethylpentane, PRF B&J Brand, meets requirements of ASTM D2699 and D2700, Burdick & Jackson™
Supplier: Honeywell Chemicals
Cyclopentane, extra dry 95+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Isobutanol 99+% for HPLC
Supplier: Thermo Fisher Scientific