1-(oxetan-3-yl)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 1 * 1 g
Catalog # PHBLPBOX0116-1G
Supplier: Pharmablock (USA)
1-(oxetan-3-yl)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 1 * 1 g
Catalog # PHBLPBOX0116-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol : Water (30:70) % (v/v), LiChrosolv®, Supelco®
Supplier: Merck
1,8-Diaminonaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
Supplier: PanReac AppliChem
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals