1-(ethanesulfonyl)azetidin-3-one 1 * 1 g
Catalog # PHBLPBLG1195-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
n-Hexane ACS, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
Supplier: PanReac AppliChem
(±)-2-Methyltetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
n-Hexane 99+% for analysis
Supplier: Thermo Fisher Scientific
FISHER CHEMICAL ACETONITRIL UHPLC GRADIENT GRAD
Supplier: MARKET SOURCE PART PROCESS
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific
Acetone, J.T.Baker®
Supplier: Avantor
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals