2-oxa-6-azaspiro[3.4]octan-5-one 1 * 1 g
N° de catalogue PHBLPBLG1182-1G
Fournisseur: Pharmablock (USA)
2-oxa-6-azaspiro[3.4]octan-5-one 1 * 1 g
N° de catalogue PHBLPBLG1182-1G
Fournisseur: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Fournisseur: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Fournisseur: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Fournisseur: Merck
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Fournisseur: VWR Chemicals
Ethanol, GPR RECTAPUR®
Fournisseur: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Fournisseur: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals