7-bromo-4-fluoro-1H-indazole 1 * 1 g
Catalog # PHBLPBJL1140-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Dodecylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific