ethyl rel-(1R,2S,4S)-4-amino-2-methyl-cyclohexanecarboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBJL072-1-1G
Supplier: Pharmablock (USA)
ethyl rel-(1R,2S,4S)-4-amino-2-methyl-cyclohexanecarboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBJL072-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99.99-100.00%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
N,N-Dimethylhydrazine
Supplier: Merck
Chloroform
Supplier: Roth Carl
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 98.5+% ACS
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% for HPLC, for gradient grade far UV
Supplier: Thermo Fisher Scientific