3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
Catalog # PHBLPBEB0396-1G
Supplier: Pharmablock (USA)
3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
Catalog # PHBLPBEB0396-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals