3-[2-[[2-(2,6-dioxo-3-piperidyl)-1,3-dioxo-isoindolin-4-yl]amino]ethoxy]propanoic acid 1 * 1 g
PHBLPBCH962-1G
3-[2-[[2-(2,6-dioxo-3-piperidyl)-1,3-dioxo-isoindolin-4-yl]amino]ethoxy]propanoic acid 1 * 1 g
PHBLPBCH962-1G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals