O1-tert-butyl O5-ethyl 4-hydroxy-3,6-dihydro-2H-pyridine-1,5-dicarboxylate 1 * 25 g
Catalog # PHBLPBB9908-25G
Supplier: Pharmablock (USA)
O1-tert-butyl O5-ethyl 4-hydroxy-3,6-dihydro-2H-pyridine-1,5-dicarboxylate 1 * 25 g
Catalog # PHBLPBB9908-25G
Supplier: Pharmablock (USA)
Frequently Bought Together
Diethyl ether stabilised Ph. Eur.
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
Supplier: PanReac AppliChem
Toluene ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS for analysis
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Pyridine 99+%, extra pure
Supplier: Thermo Fisher Scientific