N-ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 1 * 1 g
Catalog # PHBLPBB8290-1G
Supplier: Pharmablock (USA)
N-ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 1 * 1 g
Catalog # PHBLPBB8290-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Lithium perchlorate trihydrate 99%, extra pure
Supplier: Thermo Fisher Scientific
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 99+% for HPLC
Supplier: Thermo Fisher Scientific
n-Hexane, extra dry 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) 98%, pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.8% ACS
Supplier: Thermo Fisher Scientific