(2S,4R)-1-[(2S)-2-(tert-butoxycarbonylamino)non-8-enoyl]-4-hydroxy-pyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBA0619-1G
Supplier: Pharmablock (USA)
(2S,4R)-1-[(2S)-2-(tert-butoxycarbonylamino)non-8-enoyl]-4-hydroxy-pyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBA0619-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Diethylene glycol monobutyl ether, Sigma-Aldrich®
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
1,2,3,4-TETRAHYDRONAPHTHALENE FOR SYN... 1 * 200 kg
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem