(2,4-difluorophenyl)boronic acid 1 * 10 g
Catalog # PHBLPB125277-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Diisodecyl phthalate, Sigma-Aldrich®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
N-Ethyldiisopropylamine 99.5+%
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
n-Decane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol), extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Videojet Technologies, OPLOSMIDDEL 750 M
Supplier: MARKET SOURCE PART PROCESS
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Butanone (Methylethylketone) (Reag. USP, Ph. Eur.) for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
Pyridine ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether 99.9%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol 99.5+%
Supplier: Thermo Fisher Scientific