4-[2,6-difluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine 1 * 1 g
Catalog # PHBLPB112940-1G
Supplier: Pharmablock (USA)
4-[2,6-difluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine 1 * 1 g
Catalog # PHBLPB112940-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform
Supplier: Roth Carl
Benzyl alcohol, extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hydrindantin dihydrate, Supelco®
Supplier: Merck
Cyclohexane, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Supplier: Thermo Fisher Scientific
n-Heptane, anhydrous (max. 0.003% H₂O)
Supplier: VWR Chemicals
4-(Methylamino)phenol sulphate 99.0-101.5% ACS
Supplier: Thermo Fisher Scientific