5-bromo-1,3-dihydro-2-benzofuran-1,3-dione 1 * 100 g
Catalog # PHBLPB112899-100G
Supplier: Pharmablock (USA)
5-bromo-1,3-dihydro-2-benzofuran-1,3-dione 1 * 100 g
Catalog # PHBLPB112899-100G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol anhydrous, 99.5 % 1 * 2 L
Supplier: Merck
Petroleum spirit, 40…60 °C, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.9%
Supplier: Honeywell Chemicals
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Water, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99%, Reagent Grade
Supplier: Honeywell Chemicals