N,N-dimethyl-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]acetamide 1 * 1 g
Catalog # PHBLPB07328-1G
Supplier: Pharmablock (USA)
N,N-dimethyl-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]acetamide 1 * 1 g
Catalog # PHBLPB07328-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Barbituric acid, Supelco®
Supplier: Merck
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Glycerine ≥86%
Supplier: Roth Carl
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals