tert-butyl 3-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]methyl}azetidine-1-carboxylate 1 * 1 g
Catalog # PHBLPB07272-1G
Supplier: Pharmablock (USA)
tert-butyl 3-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]methyl}azetidine-1-carboxylate 1 * 1 g
Catalog # PHBLPB07272-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Heptane ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific