5-chloro-thiazole-2-carbonitrile 1 * 1 g
Catalog # PHBLPB05250-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Cyclohexane for HPLC
Supplier: Thermo Fisher Scientific
Chloroform stabilized with ethanol pure 1 * 1.000 mL
Supplier: PanReac AppliChem
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem