3,4-dichloropyridazine 1 * 1 g
Catalog # PHBLPB05235-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-Dimethoxyethane, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem