5 COMPONENTS IN N,N-DIMETHYLACETAMIDE 1 * 10 mL
Catalog # ACSDRD09701910.3
Supplier: CPACHEM
Frequently Bought Together
Methanol, SupraSolv® for GC-MS
Supplier: Merck
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
Sulphur trioxide pyridine complex for synthesis
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Pyridine, EMPLURA®, Supelco®
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid 99+% for spectroscopy
Supplier: Thermo Fisher Scientific