BALLAST OSRAM QTP-OPTIMAL 2x18-40 1 * 1 items
Catalog # TELR621562
Supplier: Azbil Telstar Technologies
BALLAST OSRAM QTP-OPTIMAL 2x18-40 1 * 1 items
Catalog # TELR621562
Supplier: Azbil Telstar Technologies
Frequently Bought Together
EMBOUT FERMANT AVEC RACCORDEMENT À QUEUE CANNELÉE 7 MM 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
OPERATING PANEL REPLACEMENT KIT 1 * 1 KIT
Supplier: IKA
Door switch 1 * 1 items
Supplier: LABCONCO
ETHERNET INTERFACE RJ45 1 * 1 items
Supplier: Thermo Scientific
NOZZLE 0.2MM 1 * 1 items
Supplier: VACUUBRAND
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Accessories for multi channel pipettes, mechanical, Research® plus
Supplier: EPPENDORF
FAN FOR RECIRCULATOR 1 * 1 items
Supplier: GRANT INSTRUMENTS
Pressure disk right for MM400 1 * 1 items
Supplier: Retsch GmbH
HSG ASSY PUMP UPPER W/BEARING 1 * 1 items
Supplier: Thermo Scientific