Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

VWR® Sample Vials

12,00 ml, Flat bottom, 19×65 mm, Clear, Yes, 15-425, PP, Black, PTFE liner, -
  216-3080
VWR® Sample Vials
  216-3080
VWR® Sample Vials
VWR® Sample Vials

12,00 ml, Flat bottom, 19×65 mm, Clear, Yes, 15-425, PP, Black, PTFE liner, -

 
SFr. 42.30
   
1 * 144 ST
 

Screw neck vials with flat bottom made from clear borosilicate glass.

  • Highly resistant against chemicals
  • High temperature resistance
  • Delivered with screw caps

Ideal for storage of liquid and powders.

 
  • Cap included:
    Yes
  • Cap material:
    PP
  • Cap Size:
    15-425
  • Ø×H:
    19×65 mm
  • Septum:
    PTFE liner
  • Sterile:
    -
  • Capacity:
    12,00 ml
  • Colour:
    Clear
  • Seq:
    05
  • Cap colour:
    Black
  • Base:
    Flat bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.