Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

VWR® Sample Vials

2,00 ml, Flat bottom, 12×35 mm, Clear, Yes, 8-425, PP, Black, PTFE liner, -
Catalog # 216-3076
Supplier:   VWR Collection
VWR® Sample Vials
Catalog # 216-3076
Supplier:  VWR Collection
VWR® Sample Vials
VWR® Sample Vials

2,00 ml, Flat bottom, 12×35 mm, Clear, Yes, 8-425, PP, Black, PTFE liner, -

Price
SFr. 29.70
Log in for availability
1 * 144 ST
Product Details & Documents

Screw neck vials with flat bottom made from clear borosilicate glass.

  • Highly resistant against chemicals
  • High temperature resistance
  • Delivered with screw caps

Ideal for storage of liquid and powders.

Specifications
  • Cap included:
    Yes
  • Cap material:
    PP
  • Cap Size:
    8-425
  • Ø×H:
    12×35 mm
  • Septum:
    PTFE liner
  • Sterile:
    -
  • Capacity:
    2,00 ml
  • Colour:
    Clear
  • Seq:
    01
  • Cap colour:
    Black
  • Base:
    Flat bottom
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.