GRINTECH NEM-060-25-10-860-S-1.5P CUSTOMIZED NEEDLE ENDOMICROSCOPE, SINGLET, DIAMETER 0.6 MM, DESIGN WAVELENGTH 860NM, GEOMETRICAL L ENGTH APPROX. 7.55 MM, NA 0.5, NON-COATED WORKING DISTANCE: OBJECT SIDE: 0.25 MM IN WATER / IMAGE SIDE: 0.1 MM IN AIR 1 * 1 ST
Catalog # MSPP100365
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together