ContactUsLinkComponent
MULTIELEMENT-STANDARDLÖSUNG 27 components: 10mg/l each of Al Ba Be Bi Cd Ca Cs Cr Co Cu Ga In Fe Pb Li Mg Mn Mo Ni K Rb Ag Na Sr Tl V Zn in HNO3 10% 1 * 100 mL
Catalog # CMPLRM037066L1
MULTIELEMENT-STANDARDLÖSUNG 27 components: 10mg/l each of Al Ba Be Bi Cd Ca Cs Cr Co Cu Ga In Fe Pb Li Mg Mn Mo Ni K Rb Ag Na Sr Tl V Zn in HNO3 10% 1 * 100 mL
Catalog # CMPLRM037066L1
Frequently Bought Together