(2R,3S,4S)-2-(hydroxymethyl)oxolane-3,4-diol 1 * 1 g
MSPPFCH3898274-1G
Frequently Bought Together
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
: Thermo Fisher Scientific
Toluene 99.85% for HPLC
: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
: Thermo Fisher Scientific
Butyl acetate ≥99%
: Thermo Fisher Scientific
n-Hexane ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
: PanReac AppliChem
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
2-Methylbutane ≥99%, Reagent Grade
: Honeywell Chemicals
ANGENE (2S)-2-[[(9H-FLUOREN-9-YLMETHOXY)CARBONYL]AMINO]OCTANOIC ACID 1 * 25 g
: MARKET SOURCE PART PROCESS