50 TABLETS DPD N° 3 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSPD350
Supplier: WATER ID PRIMELAB
Frequently Bought Together
PALADUR POWDER COLORLESS 100G/PK 1 * 100 g
Supplier: MARKET SOURCE PART PROCESS
[EN]IGG4 ISOTYPE CONTROL S228P/R409K 1 * 5 mg
Supplier: MARKET SOURCE PART PROCESS
SEKUMATIC FNZ 5L (3) NEUTRALIZER, LIQUID 1 * 1 items
Supplier: KAMMERER
pH Papers, without color scale
Supplier: MACHEREY-NAGEL
Diluid III Diff for hematology, J.T.Baker®
Supplier: Avantor
50 TABLETS ORGAPHOS-OX PHOTOMETER 1 * 50 items
Supplier: WATER ID PRIMELAB
Ferrous Ammonium Sulphate 0.1M Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
VARIO SG REAGENT SET 1 * 1 SET
Supplier: Lovibond
Nitrite VHR L, 500 ml
Supplier: Lovibond