50 TABLETS ALUMINIUM N° 2 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSPALM250
Supplier: WATER ID PRIMELAB
50 TABLETS ALUMINIUM N° 2 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSPALM250
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Acid red 52, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
J.T.Baker® Cell Lysis Solution, Biotech Reagent
Supplier: Avantor
Dodecamolybdophosphoric acid hydrate solution ready-to-use spray and plunge reagent for chromatography, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
0.1% GELATIN-BASED COATING SOLUTION- REA 0.1% GELATIN-BASED COATING SOLUTION- REA 1 * 100 mL
Supplier: MARKET SOURCE PART PROCESS
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS) ≥80%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Colour comparison disk Ammonium suitable for Cat. No. 920006 VISOCOLOR HE. 1 * 1 items
Supplier: MACHEREY-NAGEL
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
SHEATH/RINSE 1 * 10 L
Supplier: MARKET SOURCE PART PROCESS
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
FIXATIVE CAL-RITE 1GAL 1 * 1 items
Supplier: Epredia
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond
Nitrite VHR L, 500 ml
Supplier: Lovibond