10ML REAGENT GRADE METHANOL 1 * 10 mL
Catalog # WTIDPTT-METHA
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY