Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
50 DPD 1 PHOTOMETER POWDER-PILLOWS 1 * 50 items
Catalog # WTIDPPPD150
Supplier:
WATER ID PRIMELAB
Expand image
50 DPD 1 PHOTOMETER POWDER-PILLOWS 1 * 50 items
Catalog # WTIDPPPD150
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
VARIO SG REAGENT SET 1 * 1 SET
Supplier: Lovibond
Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific
Sulphur S - 1000 mg/l in H2O for ICP CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 30 mL
Supplier: CPACHEM
SODIUIM HYDROXIDE 30% SOLUTION 1 * 500 mL
Supplier: Spectrum Chemical