REAGENT KIT FOR PRIMELAB ID67(250 TESTS) 1 * 1 items
Catalog # WTIDPL67-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID67(250 TESTS) 1 * 1 items
Catalog # WTIDPL67-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Ammonia solution ∼32%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test strips, nitrate , Reflectoquant®, Supelco®
Supplier: Merck
Reference substances for GC analysis, Supelco®
Supplier: Merck