65ML PL SILICA LR 1 (162 TESTS) 1 * 65 mL
Catalog # WTIDPL65SILR1
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Teststifte Oberflächenenergie 30-44 mN/m 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
NANOCOLOR® Photometric Tube Tests Kits for Ammonium
Supplier: MACHEREY-NAGEL
[EN]IGG1 ISOTYPE CONTROL S239D/A330L/I33 1 * 5 mg
Supplier: MARKET SOURCE PART PROCESS
25 COMPONENT 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Stemgent Stemolecule All-Trans Retinoic Acid (100mg) 1 * 100 mg
Supplier: MARKET SOURCE PART PROCESS
Nitrite VHR L, 500 ml
Supplier: Lovibond
Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific
CYTOSPRING 1M DMSO 12x2ml 1 * 24 mL
Supplier: MARKET SOURCE PART PROCESS
Ammonia solution 10% (w/w) in water, extra pure
Supplier: Thermo Fisher Scientific